Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM177B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324080.1 | NP_001311009.1 | 4 | 451 | ||
NM_207468.2 | NP_997351.2 | 5 | 553 | |||
NR_136691.1 | 5 | 1057 | ||||
XM_017001279.1 | XP_016856768.1 | 4 | 995 | |||
XM_017001280.1 | XP_016856769.1 | 5 | 509 | |||
XM_017001281.1 | XP_016856770.1 | 5 | 1078 | |||
XM_017001282.1 | XP_016856771.1 | 5 | 534 | |||
XM_017001283.1 | XP_016856772.1 | 4 | 831 | |||
XM_017001284.1 | XP_016856773.1 | 5 | 424 | |||
GenBank mRNA | AK125494.1 | - | 5 | 1057 | ||
CA848039.1 | - | 4 | 403 |