Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CD46 SIRNA Assays ›
Gene Symbol:
CD46
Gene Name:
CD46 molecule
Gene Aliases:
AHUS2, MCP, MIC10, TLX, TRA2.10
Chromosome Location:
Chr.1: 207752038 - 207795516 on Build GRCh38
UniGene ID:
Hs.510402
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s533865
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CD46
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002389.4
NP_002380.3
3
527
NM_153826.3
NP_722548.1
3
527
NM_172350.2
NP_758860.1
3
527
NM_172351.2
NP_758861.1
3
527
NM_172352.2
NP_758862.1
3
527
NM_172353.2
NP_758863.1
3
527
NM_172359.2
NP_758869.1
3
527
NM_172361.2
NP_758871.1
3
527
XM_011509563.1
XP_011507865.1
3
527
XM_011509564.1
XP_011507866.1
3
527
XM_017001308.1
XP_016856797.1
3
527
XM_017001309.1
XP_016856798.1
3
527
XM_017001310.1
XP_016856799.1
3
527
GenBank mRNA
A18585.1
-
3
385
AF209712.1
-
3
352
AF209713.1
-
3
352
AF209714.1
-
3
352
AK222822.1
-
3
453
AK291227.1
-
3
481
AK296819.1
-
3
479
AU131238.2
-
3
508
BC030594.2
-
3
467
4823040
BX537451.1
-
3
498
BX640613.1
-
3
509
BX649050.1
-
3
498
D84105.1
-
3
373
DB078198.1
-
3
527
EF076055.1
-
3
385
EF076056.1
-
3
385
EF076057.1
-
3
385
EF076058.1
-
3
385
M58050.1
-
3
386
S51940.1
-
3
439
X59405.1
-
3
497
X59406.1
-
3
456
X59407.1
-
3
456
X59408.1
-
3
497
X59409.1
-
3
456
X59410.1
-
3
456
Y00651.1
-
3
395
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
adaptive immune response
T cell mediated immunity
proteolysis
complement activation, classical pathway
single fertilization
regulation of Notch signaling pathway
positive regulation of gene expression
negative regulation of gene expression
regulation of complement activation
interleukin-10 production
positive regulation of interleukin-10 production
sequestering of extracellular ligand from receptor
positive regulation of T cell proliferation
negative regulation of catalytic activity
positive regulation of memory T cell differentiation
innate immune response
positive regulation of regulatory T cell differentiation
negative regulation of complement activation
viral entry into host cell
positive regulation of transforming growth factor beta production
virus receptor activity
complement binding
endopeptidase activity
enzyme inhibitor activity
receptor activity
protein binding
cadherin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0