Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244644.1 | NP_001231573.1 | 4 | 671 | ||
NM_014043.3 | NP_054762.2 | 5 | 763 | |||
XM_011533576.2 | XP_011531878.1 | 5 | 594 | |||
GenBank mRNA | AF151842.1 | - | 5 | 722 | ||
AK002165.1 | - | 5 | 736 | |||
AK002180.1 | - | 5 | 741 | |||
AK222654.1 | - | 5 | 723 | |||
AK296072.1 | - | 4 | 663 | |||
AK316585.1 | - | 5 | 727 | |||
AL080122.1 | - | 5 | 736 | |||
BC001553.1 | - | 5 | 579 | 3460712 | ||
CR533456.1 | - | 5 | 503 |