Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPHP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291593.1 | NP_001278522.1 | 27 | 4218 | ||
NM_001291594.1 | NP_001278523.1 | 26 | 4041 | |||
NM_015102.4 | NP_055917.1 | 30 | 4528 | |||
NR_111987.1 | 33 | 5075 | ||||
XM_006710563.3 | XP_006710626.1 | 30 | 4374 | |||
XM_011541213.1 | XP_011539515.1 | 29 | 4295 | |||
XM_011541214.1 | XP_011539516.1 | 29 | 4256 | |||
XM_011541215.1 | XP_011539517.1 | 28 | 4187 | |||
XM_011541216.2 | XP_011539518.1 | 31 | 5002 | |||
XM_011541217.2 | XP_011539519.1 | 30 | 4866 | |||
XM_011541218.2 | XP_011539520.1 | 31 | 4535 | |||
XM_017000996.1 | XP_016856485.1 | 29 | 4253 | |||
XM_017000997.1 | XP_016856486.1 | 30 | 4797 | |||
XM_017000999.1 | XP_016856488.1 | 30 | 4293 | |||
XM_017001000.1 | XP_016856489.1 | 29 | 4126 | |||
XM_017001001.1 | XP_016856490.1 | 24 | 3565 | |||
XM_017001003.1 | XP_016856492.1 | 22 | 3356 | |||
GenBank mRNA | AB014573.1 | - | 25 | 3628 | ||
AF537130.1 | - | 30 | 4526 | |||
AK124074.1 | - | 26 | 3994 | |||
AK127333.1 | - | 3 | 3895 | |||
AK314326.1 | - | 27 | 4192 | |||
AY118228.1 | - | 30 | 4463 | |||
BC040520.1 | - | 33 | 5076 | 5295140 | ||
BC050076.1 | - | 3 | 3489 |