Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PYGM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164716.1 | NP_001158188.1 | 5, 6 | 1407 | ||
GenBank mRNA | AF066859.1 | - | 7, 8 | 854 | ||
AK056607.1 | - | 5, 6 | 666 | |||
AK299461.1 | - | 7, 8 | 873 | |||
AK314474.1 | - | 7, 8 | 858 | |||
BC029305.1 | - | 7, 8 | 903 | |||
BC126392.1 | - | 7, 8 | 1253 | 8992108 | ||
BC130514.1 | - | 7, 8 | 1253 | 40146545 |