Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other WNK1 SIRNA Assays ›
Gene Symbol:
WNK1
Gene Name:
WNK lysine deficient protein kinase 1
Gene Aliases:
HSAN2, HSN2, KDP, PPP1R167, PRKWNK1, PSK, p65
Chromosome Location:
Chr.12: 752552 - 911452 on Build GRCh38
UniGene ID:
Hs.744906
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s534025
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: WNK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001184985.1
NP_001171914.1
6
2249
NM_014823.2
NP_055638.2
6
2249
NM_018979.3
NP_061852.3
6
2249
NM_213655.4
NP_998820.3
6
2249
XM_006719003.2
XP_006719066.1
6
2574
XM_011520997.2
XP_011519299.1
6
2620
XM_011520998.2
XP_011519300.1
6
2574
XM_011520999.2
XP_011519301.1
6
2574
XM_011521000.2
XP_011519302.1
6
2574
XM_011521001.2
XP_011519303.1
6
2574
XM_011521002.2
XP_011519304.1
6
2574
XM_011521003.2
XP_011519305.1
6
2574
XM_011521004.2
XP_011519306.1
6
2574
XM_011521005.2
XP_011519307.1
3
1058
XM_011521006.2
XP_011519308.1
6
2574
XM_011521007.2
XP_011519309.1
6
2574
XM_011521008.2
XP_011519310.1
6
2574
XM_011521009.2
XP_011519311.1
6
2574
XM_017019834.1
XP_016875323.1
6
2574
XM_017019835.1
XP_016875324.1
6
2574
XM_017019836.1
XP_016875325.1
6
2574
XM_017019837.1
XP_016875326.1
6
2574
XM_017019838.1
XP_016875327.1
6
2574
XM_017019839.1
XP_016875328.1
6
2574
GenBank mRNA
AB002342.2
-
6
1404
AF061944.1
-
6
2249
AJ296290.1
-
6
1606
JQ358908.1
-
6
1606
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of systemic arterial blood pressure
protein phosphorylation
ion transport
negative regulation of phosphatase activity
peptidyl-threonine phosphorylation
positive regulation of ion transmembrane transporter activity
intracellular signal transduction
protein autophosphorylation
neuron development
regulation of cellular process
negative regulation of protein serine/threonine kinase activity
negative regulation of pancreatic juice secretion
regulation of ion homeostasis
magnesium ion binding
protein serine/threonine kinase activity
protein binding
ATP binding
chloride channel inhibitor activity
potassium channel inhibitor activity
protein kinase binding
phosphatase binding
protein serine/threonine kinase inhibitor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-j4kkz:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0