Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000195.4 | NP_000186.2 | 5 | 593 | ||
NM_001311345.1 | NP_001298274.1 | 5 | 602 | |||
NM_001322476.1 | NP_001309405.1 | 5 | 522 | |||
NM_001322477.1 | NP_001309406.1 | 5 | 602 | |||
NM_001322478.1 | NP_001309407.1 | 5 | 522 | |||
NM_001322479.1 | NP_001309408.1 | 5 | 602 | |||
NM_001322487.1 | NP_001309416.1 | 5 | 602 | |||
NM_001322490.1 | NP_001309419.1 | 5 | 602 | |||
NM_001322491.1 | NP_001309420.1 | 5 | 522 | |||
NM_001322492.1 | NP_001309421.1 | 5 | 522 | |||
NM_182639.3 | NP_872577.1 | 5 | 602 | |||
XM_005269757.4 | XP_005269814.1 | 5 | 535 | |||
XM_017016173.1 | XP_016871662.1 | 5 | 600 | |||
GenBank mRNA | AK295821.1 | - | 5 | 558 | ||
BC000175.2 | - | 5 | 552 | |||
DA479757.1 | - | 5 | 522 | |||
KU178114.1 | - | 3 | 331 | |||
KU178115.1 | - | 3 | 331 | |||
KU178116.1 | - | 3 | 331 | |||
KU178118.1 | - | 3 | 331 | |||
KU178120.1 | - | 3 | 331 | |||
U65676.1 | - | 5 | 537 | |||
U96721.1 | - | 5 | 565 |