Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000195.4 | NP_000186.2 | 19, 20 | 2194 | ||
NM_001311345.1 | NP_001298274.1 | 18, 19 | 2147 | |||
NM_001322476.1 | NP_001309405.1 | 19, 20 | 2123 | |||
NM_001322477.1 | NP_001309406.1 | 19, 20 | 2203 | |||
NM_001322478.1 | NP_001309407.1 | 18, 19 | 2024 | |||
NM_001322479.1 | NP_001309408.1 | 18, 19 | 2104 | |||
NM_001322480.1 | NP_001309409.1 | 17, 18 | 1862 | |||
NM_001322481.1 | NP_001309410.1 | 17, 18 | 1942 | |||
NM_001322482.1 | NP_001309411.1 | 16, 17 | 1843 | |||
NM_001322483.1 | NP_001309412.1 | 18, 19 | 1980 | |||
NM_001322484.1 | NP_001309413.1 | 18, 19 | 2060 | |||
NM_001322485.1 | NP_001309414.1 | 17, 18 | 1952 | |||
NM_001322487.1 | NP_001309416.1 | 19, 20 | 2246 | |||
NM_001322489.1 | NP_001309418.1 | 17, 18 | 2004 | |||
XM_005269757.4 | XP_005269814.1 | 19, 20 | 2136 | |||
XM_017016170.1 | XP_016871659.1 | 16, 17 | 1750 | |||
XM_017016171.1 | XP_016871660.1 | 17, 18 | 1957 | |||
XM_017016172.1 | XP_016871661.1 | 16, 17 | 1768 | |||
GenBank mRNA | AL833734.1 | - | 8, 9 | 846 | ||
U65676.1 | - | 19, 20 | 2138 |