Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017524.2 | NP_001017524.1 | 7 | 1829 | ||
NM_012239.5 | NP_036371.1 | 7 | 1975 | |||
XM_005252835.1 | XP_005252892.1 | 7 | 2029 | |||
XM_011519956.1 | XP_011518258.1 | 7 | 2821 | |||
XM_011519957.1 | XP_011518259.1 | 7 | 2650 | |||
XM_017017428.1 | XP_016872917.1 | 7 | 1875 | |||
XM_017017429.1 | XP_016872918.1 | 7 | 2049 | |||
XM_017017430.1 | XP_016872919.1 | 7 | 1989 | |||
XM_017017431.1 | XP_016872920.1 | 7 | 1774 | |||
GenBank mRNA | BM709066.1 | - | 1 | 196 | ||
DR762907.1 | - | 1 | 689 |