Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174150.1 | NP_001167621.1 | 7, 8 | 1315 | ||
NM_001174151.1 | NP_001167622.1 | 6, 7 | 1244 | |||
NM_001321328.1 | NP_001308257.1 | 8, 9 | 1437 | |||
NM_144996.3 | NP_659433.2 | 5, 6 | 994 | |||
NM_182896.2 | NP_878899.1 | 7, 8 | 1315 | |||
NR_033427.1 | 6, 7 | 1070 | ||||
NR_135621.1 | 6, 7 | 1061 | ||||
XM_006713532.3 | XP_006713595.1 | 8, 9 | 1434 | |||
XM_011512532.2 | XP_011510834.1 | 8, 9 | 1343 | |||
XM_011512533.2 | XP_011510835.1 | 9, 10 | 1151 | |||
XM_011512534.2 | XP_011510836.1 | 9, 10 | 1469 | |||
XM_011512535.2 | XP_011510837.1 | 8, 9 | 1328 | |||
XM_017005853.1 | XP_016861342.1 | 8, 9 | 1345 | |||
GenBank mRNA | AK296994.1 | - | 7, 8 | 1293 | ||
AK299284.1 | - | 6, 7 | 1054 | |||
AL713789.1 | - | 6, 7 | 1187 | |||
BC094725.1 | - | 5, 6 | 989 | 4818884 | ||
BC104035.1 | - | 7, 8 | 1305 | 40026398 | ||
BC104036.1 | - | 7, 8 | 1305 | 40026399 | ||
CR936753.1 | - | 5, 6 | 992 | |||
CR936756.1 | - | 6, 7 | 949 | |||
CR936763.1 | - | 8, 9 | 1389 |