Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174150.1 | NP_001167621.1 | 10 | 3304 | ||
NM_001174151.1 | NP_001167622.1 | 9 | 3233 | |||
NM_001321328.1 | NP_001308257.1 | 11 | 3426 | |||
NM_144996.3 | NP_659433.2 | 8 | 2983 | |||
NM_182896.2 | NP_878899.1 | 11 | 3148 | |||
NR_033427.1 | 9 | 3059 | ||||
NR_135621.1 | 9 | 3050 | ||||
XM_006713532.3 | XP_006713595.1 | 11 | 3423 | |||
XM_011512532.2 | XP_011510834.1 | 11 | 3332 | |||
XM_011512533.2 | XP_011510835.1 | 12 | 3140 | |||
XM_011512534.2 | XP_011510836.1 | 12 | 3458 | |||
XM_011512535.2 | XP_011510837.1 | 11 | 3317 | |||
XM_017005853.1 | XP_016861342.1 | 11 | 3334 | |||
GenBank mRNA | AL713789.1 | - | 9 | 3176 | ||
BC094725.1 | - | 8 | 2978 | 4818884 | ||
CR936753.1 | - | 8 | 2981 | |||
CR936756.1 | - | 9 | 2938 |