Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113378.1 | NP_001106849.1 | 23 | 2443 | ||
NM_018193.2 | NP_060663.2 | 23 | 2443 | |||
XM_011521756.2 | XP_011520058.1 | 23 | 2467 | |||
XM_011521757.2 | XP_011520059.1 | 23 | 2463 | |||
XM_011521764.2 | XP_011520066.1 | 23 | 2467 | |||
XM_011521767.2 | XP_011520069.1 | 23 | 2437 | |||
GenBank mRNA | AB058697.1 | - | 9 | 2367 | ||
AK001581.1 | - | 17 | 1832 | |||
AK055176.1 | - | 23 | 2402 | |||
BC140769.1 | - | 23 | 2400 | 9021637 | ||
BC144483.1 | - | 23 | 2400 | 9053010 | ||
BQ050477.1 | - | 5 | 487 | |||
EF469766.1 | - | 22 | 2353 | |||
EF567077.1 | - | 22 | 2353 |