Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321134.1 | NP_001308063.1 | 4, 5 | 461 | ||
XM_005257844.3 | XP_005257901.1 | 4, 5 | 422 | |||
XM_017025386.1 | XP_016880875.1 | 4, 5 | 422 | |||
XM_017025387.1 | XP_016880876.1 | 4, 5 | 419 | |||
XM_017025392.1 | XP_016880881.1 | 4, 5 | 422 | |||
XM_017025393.1 | XP_016880882.1 | 4, 5 | 419 | |||
XM_017025394.1 | XP_016880883.1 | 4, 5 | 770 | |||
XM_017025395.1 | XP_016880884.1 | 4, 5 | 770 | |||
GenBank mRNA | CD048223.1 | - | 4, 5 | 438 |