Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MC4R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005912.2 | NP_005903.2 | 1 | 1274 | ||
GenBank mRNA | AK314130.1 | - | 1 | 1274 | ||
BC069172.1 | - | 1 | 855 | 7216994 | ||
BC101802.1 | - | 1 | 915 | 8069308 | ||
BC111992.1 | - | 1 | 915 | 8327460 |
Set Membership: |
![]() |