Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNRHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000406.2 | NP_000397.1 | 3 | 2631 | ||
NM_001012763.1 | NP_001012781.1 | 3 | 2503 | |||
GenBank mRNA | AY392011.1 | - | 3 | 880 | ||
BC113546.1 | - | 3 | 906 | 8322598 | ||
L03380.1 | - | 3 | 935 | |||
L07949.1 | - | 3 | 904 | |||
S60587.1 | - | 3 | 904 | |||
S77472.1 | - | 3 | 905 | |||
Z81148.1 | - | 3 | 752 |
Set Membership: |
![]() |