Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204813.1 | NP_001191742.1 | 4 | 1437 | ||
NM_002526.3 | NP_002517.1 | 4 | 1437 | |||
GenBank mRNA | AK075008.1 | - | 4 | 912 | ||
AK075083.1 | - | 1 | 139 | |||
AK314661.1 | - | 4 | 951 | |||
BC065937.1 | - | 4 | 906 | 6163101 | ||
DQ186653.1 | - | 4 | 900 | |||
X55740.1 | - | 4 | 930 |