Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184728.2 | NP_001171657.1 | 6, 7 | 796 | ||
NM_001184729.2 | NP_001171658.1 | 7, 8 | 964 | |||
NM_001184730.2 | NP_001171659.1 | 5, 6 | 658 | |||
NM_015937.5 | NP_057021.2 | 7, 8 | 964 | |||
NR_047691.1 | 7, 8 | 924 | ||||
NR_047692.1 | 6, 7 | 867 | ||||
NR_047693.1 | 6, 7 | 863 | ||||
NR_047694.1 | 6, 7 | 786 | ||||
NR_047695.1 | 4, 5 | 557 | ||||
XM_005260430.2 | XP_005260487.1 | 5, 6 | 636 | |||
XM_005260432.2 | XP_005260489.1 | 6, 7 | 758 | |||
GenBank mRNA | AB057724.1 | - | 7, 8 | 874 | ||
AF132940.1 | - | 7, 8 | 868 | |||
AK075469.1 | - | 7, 8 | 888 | |||
AK225517.1 | - | 7, 8 | 894 | |||
AK293294.1 | - | 6, 7 | 787 | |||
AK293686.1 | - | 6, 7 | 709 | |||
AK296063.1 | - | 6, 7 | 793 | |||
AK296139.1 | - | 6, 7 | 719 | |||
AK297830.1 | - | 3 | 443 | |||
AK299013.1 | - | 4, 5 | 483 | |||
AK301815.1 | - | 6, 7 | 761 | |||
AK302093.1 | - | 5, 6 | 616 | |||
AK311100.1 | - | 6, 7 | 746 | |||
AK311416.1 | - | 5, 6 | 608 | |||
AK311492.1 | - | 7, 8 | 859 | |||
AK311520.1 | - | 5, 6 | 660 | |||
AK311522.1 | - | 6, 7 | 775 | |||
AK316358.1 | - | 7, 8 | 850 | |||
AL121742.1 | - | 7, 8 | 867 | |||
AY358588.1 | - | 7, 8 | 871 | |||
BC015022.1 | - | 7, 8 | 893 | 3921680 | ||
BC110892.1 | - | 7, 8 | 872 | |||
BC136827.1 | - | 7, 8 | 902 | 9020818 | ||
BC136828.1 | - | 7, 8 | 902 | 9020819 | ||
BX537612.1 | - | 5, 6 | 2302 |