Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184728.2 | NP_001171657.1 | 11 | 2049 | ||
NM_001184729.2 | NP_001171658.1 | 11 | 2016 | |||
NM_001184730.2 | NP_001171659.1 | 10 | 1911 | |||
NM_015937.5 | NP_057021.2 | 12 | 2217 | |||
NR_047691.1 | 12 | 2177 | ||||
NR_047692.1 | 11 | 2120 | ||||
NR_047693.1 | 11 | 2116 | ||||
NR_047694.1 | 11 | 2039 | ||||
NR_047695.1 | 9 | 1810 | ||||
XM_005260430.2 | XP_005260487.1 | 9 | 1688 | |||
XM_005260432.2 | XP_005260489.1 | 11 | 2011 | |||
GenBank mRNA | AF132940.1 | - | 12 | 2121 | ||
AK075469.1 | - | 12 | 2141 | |||
AK225517.1 | - | 11 | 1946 | |||
AK296139.1 | - | 11 | 1972 | |||
AL121742.1 | - | 12 | 2120 | |||
AY358588.1 | - | 12 | 2124 | |||
BC015022.1 | - | 12 | 2146 | 3921680 | ||
BC110892.1 | - | 12 | 2125 | |||
BP392024.1 | - | 1 | 325 | |||
BX537612.1 | - | 9 | 3354 |