Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000073.2 | NP_000064.1 | 4 | 437 | ||
XM_005271724.3 | XP_005271781.1 | 4 | 474 | |||
XM_006718941.2 | XP_006719004.1 | 4 | 425 | |||
GenBank mRNA | AK223314.1 | - | 4 | 390 | ||
AK310578.1 | - | 5 | 565 | |||
AK310622.1 | - | 5 | 595 | |||
AK313966.1 | - | 4 | 425 | |||
AK315895.1 | - | 4 | 381 | |||
BC113830.1 | - | 4 | 383 | 40029587 | ||
X04145.1 | - | 4 | 394 |