Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PURA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005859.4 | NP_005850.1 | 1 | 1439 | ||
GenBank mRNA | BC036087.1 | - | 1 | 1392 | 5284976 | |
X91648.1 | - | 1 | 413 |