Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252020.1 | NP_001238949.1 | 3 | 589 | ||
NM_001252024.1 | NP_001238953.1 | 4 | 377 | |||
NM_001252030.1 | NP_001238959.1 | 3 | 291 | |||
NM_002420.5 | NP_002411.3 | 3 | 291 | |||
GenBank mRNA | AF071787.1 | - | 3 | 268 | ||
BC005892.1 | - | 3 | 286 | |||
BC017849.1 | - | 3 | 286 | |||
BC058286.1 | - | 3 | 295 | |||
BU933033.1 | - | 3 | 294 | |||
HM135790.1 | - | 4 | 372 | |||
HM135791.1 | - | 3 | 589 |