Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000434.3 | NP_000425.1 | 3 | 593 | ||
GenBank mRNA | AB209011.1 | - | 2 | 1005 | ||
AF040958.1 | - | 3 | 566 | |||
AK290966.1 | - | 3 | 481 | |||
AK313006.1 | - | 3 | 480 | |||
BC000722.2 | - | 3 | 534 | 3506824 | ||
BC011900.2 | - | 3 | 465 | 4101695 | ||
BT007206.1 | - | 3 | 437 | |||
CR456717.1 | - | 3 | 437 | |||
CR541916.1 | - | 3 | 437 | |||
U84246.1 | - | 3 | 491 | |||
X78687.1 | - | 3 | 503 |