Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIDEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199551.1 | NP_001186480.1 | 7 | 1204 | ||
NM_001199552.1 | NP_001186481.1 | 6 | 1174 | |||
NM_001199623.1 | NP_001186552.1 | 6 | 1177 | |||
NM_001321142.1 | NP_001308071.1 | 7 | 1287 | |||
NM_001321143.1 | NP_001308072.1 | 6 | 1133 | |||
NM_001321144.1 | NP_001308073.1 | 5 | 1020 | |||
NM_022094.3 | NP_071377.2 | 6 | 1258 | |||
GenBank mRNA | AF303893.1 | - | 6 | 1258 | ||
AK024524.1 | - | 5 | 1017 | |||
AK024530.1 | - | 5 | 1017 | |||
AY364640.1 | - | 5 | 1104 | |||
BC016851.1 | - | 5 | 1153 | 3851667 |
Gene Symbol: CIDECP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF230335.1 | - | 3 | 1060 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK096121.1 | - | 2 | 3032 | Chr. 3 - 10017491 - 10021842 [-] on Build GRCh38 |