Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIDEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199551.1 | NP_001186480.1 | 7 | 1179 | ||
NM_001199552.1 | NP_001186481.1 | 6 | 1149 | |||
NM_001199623.1 | NP_001186552.1 | 6 | 1152 | |||
NM_001321142.1 | NP_001308071.1 | 7 | 1262 | |||
NM_001321143.1 | NP_001308072.1 | 6 | 1108 | |||
NM_001321144.1 | NP_001308073.1 | 5 | 995 | |||
NM_022094.3 | NP_071377.2 | 6 | 1233 | |||
GenBank mRNA | AF303893.1 | - | 6 | 1233 | ||
AK024524.1 | - | 5 | 992 | |||
AK024530.1 | - | 5 | 992 | |||
AY364640.1 | - | 5 | 1079 | |||
BC016851.1 | - | 5 | 1128 | 3851667 |