Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIDEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199551.1 | NP_001186480.1 | 7 | 858 | ||
NM_001199552.1 | NP_001186481.1 | 6 | 828 | |||
NM_001199623.1 | NP_001186552.1 | 6 | 831 | |||
NM_001321142.1 | NP_001308071.1 | 7 | 941 | |||
NM_001321143.1 | NP_001308072.1 | 6 | 787 | |||
NM_001321144.1 | NP_001308073.1 | 5 | 674 | |||
NM_022094.3 | NP_071377.2 | 6 | 912 | |||
GenBank mRNA | AF303893.1 | - | 6 | 912 | ||
AK024524.1 | - | 5 | 671 | |||
AK024530.1 | - | 5 | 671 | |||
AY364638.1 | - | 3 | 427 | |||
AY364640.1 | - | 5 | 758 | |||
BC016851.1 | - | 5 | 807 | 3851667 | ||
CD519587.1 | - | 5 | 807 |