Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PAK3 SIRNA Assays ›
Gene Symbol:
PAK3
Gene Name:
p21 (RAC1) activated kinase 3
Gene Aliases:
ARA, MRX30, MRX47, OPHN3, PAK-3, PAK3beta, bPAK, beta-PAK
Chromosome Location:
Chr.X: 110944241 - 111227361 on Build GRCh38
UniGene ID:
Hs.593599
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s534741
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PAK3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128166.2
NP_001121638.1
3
540
NM_001128167.2
NP_001121639.1
2
467
NM_001128168.2
NP_001121640.1
5
681
NM_001128172.2
NP_001121644.1
1
79
NM_001128173.2
NP_001121645.1
5
681
NM_001324325.1
NP_001311254.1
3
618
NM_001324326.1
NP_001311255.1
3
463
NM_001324327.1
NP_001311256.1
4
530
NM_001324328.1
NP_001311257.1
3
463
NM_001324329.1
NP_001311258.1
5
678
NM_001324330.1
NP_001311259.1
3
615
NM_001324331.1
NP_001311260.1
4
530
NM_001324332.1
NP_001311261.1
5
681
NM_001324333.1
NP_001311262.1
4
611
NM_001324334.1
NP_001311263.1
4
611
NM_002578.4
NP_002569.1
5
678
NR_136740.1
3
463
NR_136741.1
4
530
NR_136742.1
5
678
NR_136743.1
4
614
NR_136744.1
5
681
NR_136745.1
5
681
NR_136746.1
6
873
NR_136747.1
4
530
NR_136748.1
2
227
XM_005262132.2
XP_005262189.1
5
673
XM_005262133.3
XP_005262190.1
5
673
XM_006724654.2
XP_006724717.1
4
520
XM_006724655.2
XP_006724718.1
3
463
XM_011530962.1
XP_011529264.1
5
671
XM_011530967.2
XP_011529269.1
4
611
XM_011530968.2
XP_011529270.1
5
687
XM_011530969.1
XP_011529271.1
3
532
XM_011530971.2
XP_011529273.1
5
673
XM_017029557.1
XP_016885046.1
3
532
XM_017029558.1
XP_016885047.1
4
520
XM_017029559.1
XP_016885048.1
3
532
XM_017029560.1
XP_016885049.1
3
463
XM_017029561.1
XP_016885050.1
5
680
XM_017029562.1
XP_016885051.1
5
673
XM_017029563.1
XP_016885052.1
5
673
GenBank mRNA
AB102659.1
-
1
52
AF068864.1
-
1
52
AF155651.1
-
5
443
AK128022.1
-
4
766
AK290504.1
-
2
467
AK315284.1
-
3
384
AM943850.1
-
1
52
AM943851.1
-
1
52
AM943852.1
-
1
52
BC117353.1
-
2
146
BC117355.1
-
2
146
BC143544.1
-
2
146
BQ268758.1
-
5
494
DB053586.1
-
3
466
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPK activity
mitotic cell cycle
stimulatory C-type lectin receptor signaling pathway
axonogenesis
dendrite development
regulation of actin filament polymerization
T cell costimulation
positive regulation of neuron apoptotic process
ephrin receptor signaling pathway
synapse organization
T cell receptor signaling pathway
dendritic spine morphogenesis
positive regulation of dendritic spine morphogenesis
cellular response to organic cyclic compound
positive regulation of DNA biosynthetic process
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
MAP kinase kinase activity
protein binding
ATP binding
Rho GTPase binding
SH3 domain binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-7cf5v:80/10.239.163.60:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0