Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DMD SIRNA Assays ›
Gene Symbol:
DMD
Gene Name:
dystrophin
Gene Aliases:
BMD, CMD3B, DXS142, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, MRX85
Chromosome Location:
Chr.X: 31119219 - 33339609 on Build GRCh38
UniGene ID:
Hs.495912
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s534743
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DMD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000109.3
NP_000100.2
76
11168
NM_004006.2
NP_003997.1
76
11092
NM_004009.3
NP_004000.1
76
11099
NM_004010.3
NP_004001.1
76
11182
NM_004011.3
NP_004002.2
48
6870
NM_004012.3
NP_004003.1
48
7013
NM_004013.2
NP_004004.1
33
4509
NM_004014.2
NP_004005.1
22
2722
NM_004015.2
NP_004006.1
15
1722
NM_004016.2
NP_004007.1
15
1722
NM_004017.2
NP_004008.1
14
1683
NM_004018.2
NP_004009.1
14
1683
NM_004020.3
NP_004011.2
29
4179
NM_004021.2
NP_004012.1
33
4509
NM_004022.2
NP_004013.1
32
4470
NM_004023.2
NP_004014.1
29
4179
XM_006724468.2
XP_006724531.1
76
11045
XM_006724469.3
XP_006724532.1
76
11019
XM_006724470.3
XP_006724533.1
75
11006
XM_006724473.2
XP_006724536.1
75
10907
XM_006724474.3
XP_006724537.1
72
10715
XM_006724475.2
XP_006724538.1
72
10715
XM_011545467.1
XP_011543769.1
75
10922
XM_017029328.1
XP_016884817.1
75
11006
XM_017029331.1
XP_016884820.1
36
5621
GenBank mRNA
AB208836.1
-
29
3917
AK299936.1
-
11
1434
BC028720.1
-
15
1773
BC070078.1
-
14
1684
BC094758.1
-
15
1736
BC127103.1
-
30
4033
BC150141.1
-
30
4033
KU178030.1
-
15
1644
KU178031.1
-
15
1644
KU178032.1
-
11
1314
M18533.1
-
76
11056
M92650.1
-
14
1657
X14298.1
-
76
10946
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
regulation of heart rate
cytoskeleton organization
muscle organ development
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
regulation of skeletal muscle contraction by regulation of release of sequestered calcium ion
regulation of skeletal muscle contraction
muscle attachment
muscle filament sliding
negative regulation of peptidyl-serine phosphorylation
cellular protein localization
response to muscle stretch
peptide biosynthetic process
cellular protein complex assembly
muscle cell cellular homeostasis
muscle fiber development
cardiac muscle contraction
regulation of ryanodine-sensitive calcium-release channel activity
cardiac muscle cell action potential
regulation of voltage-gated calcium channel activity
negative regulation of peptidyl-cysteine S-nitrosylation
positive regulation of sodium ion transmembrane transporter activity
dystroglycan binding
actin binding
structural constituent of cytoskeleton
protein binding
zinc ion binding
structural constituent of muscle
myosin binding
vinculin binding
nitric-oxide synthase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-l9dfq:80/10.239.161.188:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0