Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR2A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005328.1 | NP_001005328.1 | 1 | 872 | ||
GenBank mRNA | BC136704.1 | - | 1 | 966 | 9020694 | |
BC136711.1 | - | 1 | 966 | 9020701 |
Gene Symbol: OR2A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030908.2 | NP_112170.1 | 1 | 872 | ||
GenBank mRNA | BC120953.2 | - | 1 | 872 | 40115181 | |
BC120954.1 | - | 1 | 872 | 40115185 | ||
BI020419.1 | - | 1 | 240 |