Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101676.1 | NP_001095146.1 | 4 | 576 | ||
NM_207506.2 | NP_997389.2 | 4 | 576 | |||
NR_109794.1 | 4 | 491 | ||||
XM_011517031.2 | XP_011515333.1 | 4 | 616 | |||
XM_011517034.2 | XP_011515336.1 | 4 | 615 | |||
XM_011517035.2 | XP_011515337.1 | 4 | 615 | |||
XM_017013409.1 | XP_016868898.1 | 4 | 588 | |||
XM_017013410.1 | XP_016868899.1 | 4 | 616 | |||
XM_017013411.1 | XP_016868900.1 | 4 | 586 | |||
GenBank mRNA | AK096777.1 | - | 4 | 512 | ||
AK311704.1 | - | 4 | 491 | |||
BC121818.1 | - | 4 | 509 | 40109130 | ||
BC124554.1 | - | 4 | 509 | 40109129 | ||
DA335826.1 | - | 4 | 576 | |||
DA459013.1 | - | 4 | 555 | |||
DB228621.1 | - | 4 | 491 |