Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142459.1 | NP_001135931.2 | 6 | 1908 | ||
NM_001142460.1 | NP_001135932.2 | 5 | 1794 | |||
NM_080871.3 | NP_543147.2 | 6 | 1712 | |||
GenBank mRNA | AF417920.1 | - | 5 | 1796 | ||
AK055536.1 | - | 6 | 1618 | |||
AK125565.1 | - | 6 | 1712 |