Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004525.2 | NP_004516.2 | 48 | 9188 | ||
XM_011511183.2 | XP_011509485.1 | 48 | 9189 | |||
XM_011511184.2 | XP_011509486.1 | 33 | 6701 | |||
GenBank mRNA | AY265357.1 | - | 48 | 9042 | ||
AY265358.1 | - | 48 | 9039 | |||
U04441.1 | - | 6 | 866 | |||
U33837.1 | - | 48 | 9079 |