Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252678.1 | NP_001239607.1 | 8 | 627 | ||
NM_001320665.1 | NP_001307594.1 | 9 | 674 | |||
NM_033028.4 | NP_149017.2 | 9 | 674 | |||
NR_045565.1 | 10 | 732 | ||||
NR_045566.1 | 9 | 987 | ||||
XM_011521848.1 | XP_011520150.1 | 9 | 649 | |||
XM_011521849.1 | XP_011520151.1 | 8 | 537 | |||
XM_017022450.1 | XP_016877939.1 | 9 | 955 | |||
XM_017022451.1 | XP_016877940.1 | 8 | 615 | |||
XM_017022452.1 | XP_016877941.1 | 8 | 845 | |||
XM_017022453.1 | XP_016877942.1 | 8 | 532 | |||
XM_017022454.1 | XP_016877943.1 | 7 | 482 | |||
GenBank mRNA | AF359281.1 | - | 9 | 646 | ||
AK075321.1 | - | 9 | 638 | |||
AK298922.1 | - | 10 | 704 | |||
AK302224.1 | - | 9 | 941 | |||
AK303706.1 | - | 8 | 622 | |||
AK314507.1 | - | 9 | 646 | |||
AY457143.1 | - | 9 | 746 | |||
BC008923.2 | - | 6 | 451 | |||
BC027624.2 | - | 9 | 644 | 4824276 | ||
BI562463.1 | - | 9 | 669 | |||
BX647855.1 | - | 8 | 598 | |||
DC372967.1 | - | 9 | 675 | |||
KU177936.1 | - | 9 | 625 |