Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: OFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003611.2 | NP_003602.1 | 14 | 1820 | ||
XM_005274604.2 | XP_005274661.1 | 13 | 1700 | |||
XM_005274606.3 | XP_005274663.1 | 15 | 1962 | |||
XM_011545592.2 | XP_011543894.1 | 15 | 1791 | |||
XM_011545594.2 | XP_011543896.1 | 13 | 1241 | |||
XM_011545595.1 | XP_011543897.1 | 13 | 1251 | |||
XM_011545597.1 | XP_011543899.1 | 15 | 1876 | |||
XM_011545598.1 | XP_011543900.1 | 3 | 319 | |||
XM_017029909.1 | XP_016885398.1 | 14 | 1396 | |||
XM_017029910.1 | XP_016885399.1 | 15 | 1604 | |||
XM_017029911.1 | XP_016885400.1 | 6 | 641 | |||
GenBank mRNA | AK225847.1 | - | 13 | 1630 | ||
AK290354.1 | - | 14 | 1805 | |||
AK297104.1 | - | 6 | 600 | |||
BC096344.3 | - | 13 | 1515 | 40008704 | ||
BC096345.1 | - | 14 | 1641 | |||
GU727634.1 | - | 13 | 1461 | |||
Y15164.1 | - | 14 | 1772 | |||
Y16355.1 | - | 14 | 2436 |