Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNPTAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024312.4 | NP_077288.2 | 3 | 451 | ||
XM_006719593.2 | XP_006719656.1 | 3 | 446 | |||
XM_011538731.2 | XP_011537033.1 | 3 | 447 | |||
XM_017019961.1 | XP_016875450.1 | 3 | 348 | |||
XM_017019962.1 | XP_016875451.1 | 3 | 430 | |||
GenBank mRNA | AI418963.1 | - | 3 | 435 | ||
AK056137.1 | - | 3 | 322 | |||
AM085438.1 | - | 3 | 438 | |||
AY687932.1 | - | 3 | 423 | |||
BC071687.1 | - | 3 | 399 | |||
BC131787.1 | - | 3 | 379 | 40126580 | ||
BP276948.1 | - | 3 | 451 |