Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145102.1 | NP_001138574.1 | 9 | 2293 | ||
NM_001145103.1 | NP_001138575.1 | 9 | 2104 | |||
NM_001145104.1 | NP_001138576.1 | 7 | 1737 | |||
NM_005902.3 | NP_005893.1 | 9 | 2528 | |||
XM_011521559.2 | XP_011519861.1 | 8 | 2555 | |||
XM_011521560.1 | XP_011519862.1 | 9 | 2467 | |||
GenBank mRNA | AK026690.1 | - | 2 | 1927 | ||
BC050743.1 | - | 9 | 2446 | 6094328 | ||
CR749287.1 | - | 1 | 1289 | |||
U68019.1 | - | 10 | 2259 |