Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANK1 SIRNA Assays ›
Gene Symbol:
ANK1
Gene Name:
ankyrin 1
Gene Aliases:
ANK, SPH1, SPH2
Chromosome Location:
Chr.8: 41653225 - 41896762 on Build GRCh38
UniGene ID:
Hs.654438
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s536724
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000037.3
NP_000028.3
8, 9
901
NM_001142446.1
NP_001135918.1
8, 9
1198
NM_020475.2
NP_065208.2
8, 9
901
NM_020476.2
NP_065209.2
8, 9
901
NM_020477.2
NP_065210.2
8, 9
901
XM_005273476.4
XP_005273533.1
8, 9
1175
XM_011544490.2
XP_011542792.1
9, 10
1097
XM_011544491.2
XP_011542793.1
9, 10
1096
XM_011544494.2
XP_011542796.1
9, 10
1096
XM_011544495.2
XP_011542797.1
9, 10
1095
XM_011544496.2
XP_011542798.1
9, 10
1097
XM_011544500.2
XP_011542802.1
8, 9
1003
XM_011544501.2
XP_011542803.1
8, 9
1003
XM_011544502.2
XP_011542804.1
8, 9
1003
XM_011544503.2
XP_011542805.1
9, 10
1094
XM_011544504.2
XP_011542806.1
8, 9
1003
XM_011544505.2
XP_011542807.1
8, 9
1003
XM_017013319.1
XP_016868808.1
9, 10
1096
XM_017013320.1
XP_016868809.1
9, 10
1098
XM_017013321.1
XP_016868810.1
9, 10
1041
XM_017013322.1
XP_016868811.1
8, 9
1003
XM_017013323.1
XP_016868812.1
9, 10
1017
XM_017013324.1
XP_016868813.1
8, 9
1175
XM_017013325.1
XP_016868814.1
8, 9
942
XM_017013326.1
XP_016868815.1
8, 9
935
XM_017013327.1
XP_016868816.1
9, 10
1096
XM_017013328.1
XP_016868817.1
9, 10
1096
XM_017013329.1
XP_016868818.1
8, 9
1173
GenBank mRNA
AB209418.1
-
8, 9
1198
M28880.1
-
8, 9
901
X16609.1
-
8, 9
901
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
exocytosis
ER to Golgi vesicle-mediated transport
cytoskeleton organization
signal transduction
positive regulation of organelle organization
maintenance of epithelial cell apical/basal polarity
protein targeting to plasma membrane
structural molecule activity
structural constituent of cytoskeleton
protein binding
cytoskeletal adaptor activity
enzyme binding
spectrin binding
ATPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-k7c2b:80/10.239.160.46:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0