Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001021.4 | NP_001012.1 | 2 | 165 | ||
NR_111943.1 | 1 | 371 | ||||
NR_111944.1 | 2 | 165 | ||||
GenBank mRNA | AK311951.1 | - | 2 | 73 | ||
BC009407.2 | - | 1 | 56 | 4334552 | ||
BC019899.1 | - | 1 | 61 | 3882924 | ||
BC020453.1 | - | 1 | 60 | |||
BC022370.1 | - | 2 | 78 | 4738331 | ||
BC049824.1 | - | 1 | 53 | 6212543 | ||
BC062715.1 | - | 2 | 163 | 4068842 | ||
BC070222.1 | - | 1 | 74 | 6738383 | ||
BC071928.1 | - | 1 | 64 | 5090053 | ||
BE905890.1 | - | 1 | 41 | |||
BI597659.1 | - | 1 | 371 | |||
BM017772.1 | - | 2 | 166 | |||
BM457623.1 | - | 1 | 58 | |||
DA310472.1 | - | 1 | 371 | |||
M13932.1 | - | 2 | 74 |
Gene Symbol: ATAD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011525273.2 | XP_011523575.1 | 1 | 23 |