Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000145.3 | NP_000136.2 | 7 | 682 | ||
NM_181446.2 | NP_852111.2 | 6 | 604 | |||
XM_011532733.2 | XP_011531035.1 | 7 | 670 | |||
XM_011532734.2 | XP_011531036.1 | 8 | 933 | |||
XM_011532735.2 | XP_011531037.1 | 4 | 263 | |||
XM_011532736.2 | XP_011531038.1 | 4 | 1439 | |||
XM_011532740.1 | XP_011531042.1 | 7 | 670 | |||
GenBank mRNA | AK292562.1 | - | 7 | 650 | ||
AK301899.1 | - | 4 | 333 | |||
AY429104.1 | - | 7 | 572 | |||
BC096831.1 | - | 7 | 572 | 7001973 | ||
BC118548.1 | - | 7 | 605 | 40007014 | ||
BC125270.1 | - | 6 | 527 | 40007010 | ||
JN003607.1 | - | 7 | 609 | |||
M65085.1 | - | 7 | 638 | |||
M95489.1 | - | 7 | 647 | |||
S59900.1 | - | 7 | 646 | |||
X68044.1 | - | 7 | 588 |