Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203453.3 | NP_982278.3 | 1 | 1462 | ||
GenBank mRNA | AK074672.1 | - | 1 | 1442 | ||
AK128369.1 | - | 1 | 1422 | |||
BC038108.2 | - | 1 | 1462 | 5268486 |