Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2H5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207118.2 | NP_997001.1 | 3 | 254 | ||
XM_017010862.1 | XP_016866351.1 | 2 | 216 | |||
GenBank mRNA | AJ634743.1 | - | 3 | 230 | ||
AK055106.1 | - | 3 | 249 | |||
BC056906.1 | - | 3 | 241 | 4446165 | ||
BC060317.1 | - | 2 | 206 | 4345335 | ||
CN403888.1 | - | 3 | 257 |