Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000146.3 | NP_000137.2 | 1 | 260 | ||
GenBank mRNA | AK026534.1 | - | 1 | 260 | ||
AK130191.1 | - | 1 | 260 | |||
AK130205.1 | - | 1 | 255 | |||
AK131048.1 | - | 1 | 260 | |||
AK131050.1 | - | 1 | 260 | |||
AK131053.1 | - | 1 | 274 | |||
AK307065.1 | - | 1 | 260 | |||
AK311773.1 | - | 1 | 260 | |||
AY466472.1 | - | 1 | 61 | |||
BC002991.1 | - | 1 | 228 | 2905327 | ||
BC004245.1 | - | 1 | 249 | 3615360 | ||
BC008439.1 | - | 1 | 260 | 4093482 | ||
BC008441.1 | - | 1 | 260 | |||
BC013928.1 | - | 1 | 264 | 3839120 | ||
BC016346.1 | - | 1 | 264 | 4066010 | ||
BC016354.1 | - | 1 | 264 | 4092360 | ||
BC016715.1 | - | 1 | 264 | 4067055 | ||
BC018990.2 | - | 1 | 232 | 3503710 | ||
BC021670.1 | - | 1 | 263 | 4051089 | ||
BC058820.1 | - | 1 | 253 | 4049525 | ||
BC062708.1 | - | 1 | 263 | 4774403 | ||
BX571748.1 | - | 1 | 259 | |||
CR456715.1 | - | 1 | 61 | |||
M10119.1 | - | 1 | 140 | |||
M11147.1 | - | 1 | 212 | |||
Y09188.1 | - | 1 | 238 |
Gene Symbol: SEC62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC105971.1 | - | 97 |
Gene Symbol: SLC4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF113686.1 | - | 1463 |