Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001394.3 | NP_001001394.1 | 1 | 1025 | ||
GenBank mRNA | AY083665.1 | - | 1 | 1025 | ||
BC017590.2 | - | 1 | 1025 | 4828023 | ||
BC024013.2 | - | 1 | 988 | 4826480 |
Gene Symbol: LOC100286922 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037694.1 | 1 | 1000 | |||
NR_037695.1 | 2 | 785 | ||||
NR_037696.1 | 2 | 648 |