Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLSCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011513013.2 | XP_011511315.2 | 8 | 4715 | ||
XM_017006898.1 | XP_016862387.1 | 9 | 2954 | |||
XM_017006899.1 | XP_016862388.1 | 8 | 4715 | |||
XM_017006901.1 | XP_016862390.1 | 9 | 4588 | |||
XM_017006905.1 | XP_016862394.1 | 10 | 4171 | |||
XM_017006906.1 | XP_016862395.1 | 10 | 3952 | |||
XM_017006907.1 | XP_016862396.1 | 10 | 3717 | |||
XM_017006908.1 | XP_016862397.1 | 10 | 3842 | |||
XM_017006909.1 | XP_016862398.1 | 9 | 3629 | |||
XM_017006912.1 | XP_016862401.1 | 8 | 3370 | |||
XM_017006914.1 | XP_016862403.1 | 8 | 3577 | |||
XM_017006915.1 | XP_016862404.1 | 8 | 3559 |