Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080509.2 | NP_001073978.1 | 4 | 405 | ||
XM_011520679.2 | XP_011518981.1 | 4 | 667 | |||
XM_011520680.2 | XP_011518982.1 | 4 | 645 | |||
GenBank mRNA | AK293947.1 | - | 4 | 352 | ||
AK300920.1 | - | 3 | 310 | |||
BC127660.1 | - | 2 | 171 | |||
BC127661.1 | - | 2 | 171 | |||
BC146917.1 | - | 4 | 351 | 9056845 | ||
BC146922.1 | - | 4 | 351 | 9056850 |