Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC37A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006607.2 | NP_001006608.2 | 9 | 3350 | ||
XM_011524841.2 | XP_011523143.1 | 8 | 3331 | |||
XM_011524842.2 | XP_011523144.1 | 9 | 3403 | |||
XM_011524843.2 | XP_011523145.1 | 7 | 3256 | |||
XM_011524844.2 | XP_011523146.1 | 7 | 3250 | |||
XM_011524846.2 | XP_011523148.1 | 6 | 3178 | |||
XM_011524848.2 | XP_011523150.1 | 5 | 3103 | |||
GenBank mRNA | AK091733.1 | - | 1 | 1262 | ||
AK126994.1 | - | 9 | 2032 | |||
AY386262.1 | - | 9 | 1771 | |||
BC088375.1 | - | 1 | 148 | |||
BC144427.1 | - | 9 | 1013 | |||
BC144428.1 | - | 9 | 1013 |
Gene Symbol: LRRC37A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014834.4 | NP_055649.4 | 9 | 3350 | ||
XM_005257879.4 | XP_005257936.1 | 9 | 3608 | |||
XM_005257880.4 | XP_005257937.1 | 7 | 3461 | |||
XM_005257882.4 | XP_005257939.1 | 10 | 3794 | |||
XM_006722210.3 | XP_006722273.1 | 5 | 3308 | |||
XM_006722211.3 | XP_006722274.1 | 4 | 3236 | |||
XM_011525539.2 | XP_011523841.1 | 7 | 3455 | |||
XM_017025472.1 | XP_016880961.1 | 11 | 3207 | |||
XM_017025473.1 | XP_016880962.1 | 9 | 854 | |||
GenBank mRNA | BC040501.1 | - | 9 | 3350 | 5295808 | |
CR749232.1 | - | 9 | 3351 |