Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008391.3 | NP_001008392.2 | 8 | 586 | ||
XM_011520139.2 | XP_011518441.1 | 7 | 447 | |||
GenBank mRNA | AK131470.1 | - | 8 | 569 | ||
BC062749.1 | - | 8 | 586 | |||
BU569581.1 | - | 8 | 585 |