Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015882.2 | NP_001015882.2 | 3 | 834 | ||
NR_037148.1 | 4 | 1072 | ||||
GenBank mRNA | AF447177.1 | - | 4 | 1041 | ||
AK095452.1 | - | 4 | 1055 | |||
BC048318.1 | - | 3 | 805 | 4456137 |