Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015882.2 | NP_001015882.2 | 4 | 1037 | ||
NR_037148.1 | 5 | 1275 | ||||
GenBank mRNA | AF447177.1 | - | 5 | 1244 | ||
AK095452.1 | - | 5 | 1258 | |||
BC048318.1 | - | 4 | 1008 | 4456137 |