Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ASPDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024656.2 | NP_001019827.2 | 5 | 810 | ||
NM_001114598.1 | NP_001108070.1 | 7 | 1006 | |||
GenBank mRNA | BC066359.1 | - | 5 | 810 | 5302622 |