Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM229B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033564.2 | NP_001028736.1 | 4 | 581 | ||
XM_017011174.1 | XP_016866663.1 | 4 | 426 | |||
XM_017011175.1 | XP_016866664.1 | 3 | 280 | |||
GenBank mRNA | AL049461.1 | - | 4 | 392 | ||
BC044912.2 | - | 4 | 448 | 5286266 |